Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003302.2 | NP_003293.2 | 5 | 927 | ||
GenBank mRNA | AB628085.1 | - | 5 | 694 | ||
AB628086.1 | - | 5 | 707 | |||
AB628087.1 | - | 4 | 629 | |||
AB628088.1 | - | 5 | 510 | |||
AF000974.1 | - | 5 | 916 | |||
AF025437.1 | - | 2 | 100 | |||
AJ001902.1 | - | 5 | 829 | |||
AK056773.1 | - | 5 | 821 | |||
AK291906.1 | - | 5 | 864 | |||
AK309312.1 | - | 5 | 818 | |||
BC002680.2 | - | 5 | 820 | 3610413 | ||
BC004249.1 | - | 5 | 832 | 3621877 | ||
BC004999.1 | - | 5 | 826 | 2958142 | ||
BC021540.1 | - | 5 | 832 | 3622017 | ||
BC028985.1 | - | 5 | 839 | 5122169 | ||
CR749505.1 | - | 5 | 755 |