Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003302.2 | NP_003293.2 | 5 | 953 | ||
GenBank mRNA | AB628085.1 | - | 5 | 720 | ||
AB628086.1 | - | 5 | 733 | |||
AB628087.1 | - | 4 | 655 | |||
AB628088.1 | - | 5 | 536 | |||
AF000974.1 | - | 5 | 942 | |||
AF025437.1 | - | 2 | 126 | |||
AJ001902.1 | - | 5 | 855 | |||
AK056773.1 | - | 5 | 847 | |||
AK291906.1 | - | 5 | 890 | |||
AK309312.1 | - | 5 | 844 | |||
BC002680.2 | - | 5 | 846 | 3610413 | ||
BC004249.1 | - | 5 | 858 | 3621877 | ||
BC004999.1 | - | 5 | 852 | 2958142 | ||
BC021540.1 | - | 5 | 858 | 3622017 | ||
BC028985.1 | - | 5 | 865 | 5122169 | ||
CR749505.1 | - | 5 | 781 |