Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003327.3 | NP_003318.1 | 3, 4 | 399 | ||
XM_011542074.2 | XP_011540376.1 | 1, 2 | 753 | |||
XM_011542075.2 | XP_011540377.1 | 3, 4 | 866 | |||
XM_011542076.2 | XP_011540378.1 | 1, 2 | 753 | |||
XM_017002231.1 | XP_016857720.1 | 4, 5 | 1744 | |||
XM_017002232.1 | XP_016857721.1 | 4, 5 | 1744 | |||
GenBank mRNA | BC105070.1 | - | 3, 4 | 399 | 8144073 | |
BC105072.1 | - | 3, 4 | 399 | 8144075 | ||
S76792.1 | - | 3, 4 | 349 |