Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173514.1 | NP_001166985.1 | 3, 4 | 318 | ||
NM_001173515.1 | NP_001166986.1 | 3, 4 | 315 | |||
NM_003332.3 | NP_003323.1 | 4, 5 | 351 | |||
NM_198125.2 | NP_937758.1 | 4, 5 | 348 | |||
NR_033390.1 | 3, 4 | 311 | ||||
GenBank mRNA | AF019562.1 | - | 4, 5 | 337 | ||
AJ010098.1 | - | 4, 5 | 269 | |||
AK290385.1 | - | 4, 5 | 334 | |||
AY074782.1 | - | 4, 5 | 268 | |||
BC011175.1 | - | 4, 5 | 326 | 4150408 | ||
BI908458.1 | - | 3, 4 | 283 | |||
BP295666.1 | - | 3, 4 | 284 | |||
BP302545.1 | - | 4, 5 | 351 | |||
BP305672.1 | - | 3, 4 | 302 | |||
BQ576278.1 | - | 1, 2 | 254 | |||
BT009851.1 | - | 4, 5 | 270 | |||
CR450342.1 | - | 4, 5 | 270 | |||
CR542202.1 | - | 4, 5 | 270 |