Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 4, 5 | 286 | ||
NM_003340.6 | NP_003331.1 | 4, 5 | 593 | |||
NM_181886.3 | NP_871615.1 | 4, 5 | 801 | |||
NM_181887.2 | NP_871616.1 | 4, 5 | 571 | |||
NM_181888.3 | NP_871617.1 | 4, 5 | 450 | |||
NM_181889.2 | NP_871618.1 | 4, 5 | 473 | |||
NM_181890.2 | NP_871619.1 | 4, 5 | 415 | |||
NM_181891.2 | NP_871620.1 | 4, 5 | 758 | |||
NM_181892.3 | NP_871621.1 | 3, 4 | 783 | |||
NM_181893.2 | NP_871622.1 | 3, 4 | 263 | |||
XM_017008584.1 | XP_016864073.1 | 3, 4 | 625 | |||
GenBank mRNA | AK095822.1 | - | 3, 4 | 698 | ||
AK127304.1 | - | 2, 3 | 3206 | |||
AK315775.1 | - | 4, 5 | 359 | |||
AL833414.1 | - | 4, 5 | 377 | |||
AU131013.1 | - | 4, 5 | 361 | |||
BC003395.1 | - | 4, 5 | 273 | 3447673 | ||
BC037894.2 | - | 4, 5 | 570 | 5275356 | ||
BC066917.1 | - | 4, 5 | 304 | 4792462 | ||
BG613555.1 | - | 4, 5 | 398 | |||
BM541512.1 | - | 4, 5 | 555 | |||
BU567649.1 | - | 4, 5 | 443 | |||
BU570661.1 | - | 3, 4 | 218 | |||
CR456916.1 | - | 3, 4 | 108 | |||
DA335600.1 | - | 4, 5 | 351 | |||
DA550074.1 | - | 4, 5 | 205 | |||
DA557036.1 | - | 4, 5 | 520 | |||
DA808523.1 | - | 4, 5 | 526 | |||
DA965437.1 | - | 3, 4 | 269 | |||
DB234695.1 | - | 3, 4 | 207 | |||
HY109217.1 | - | 4, 5 | 415 | |||
U39318.1 | - | 3, 4 | 153 |