Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 6 | 432 | ||
NM_003340.6 | NP_003331.1 | 6 | 739 | |||
NM_181886.3 | NP_871615.1 | 6 | 947 | |||
NM_181887.2 | NP_871616.1 | 6 | 717 | |||
NM_181888.3 | NP_871617.1 | 6 | 596 | |||
NM_181889.2 | NP_871618.1 | 6 | 619 | |||
NM_181890.2 | NP_871619.1 | 6 | 561 | |||
NM_181891.2 | NP_871620.1 | 6 | 904 | |||
NM_181892.3 | NP_871621.1 | 5 | 929 | |||
NM_181893.2 | NP_871622.1 | 5 | 409 | |||
XM_017008584.1 | XP_016864073.1 | 5 | 771 | |||
GenBank mRNA | AK095822.1 | - | 5 | 844 | ||
AK127304.1 | - | 4 | 3352 | |||
AK315775.1 | - | 6 | 505 | |||
AL833414.1 | - | 5 | 1522 | |||
AU131013.1 | - | 6 | 507 | |||
BC003395.1 | - | 6 | 419 | 3447673 | ||
BC037894.2 | - | 6 | 716 | 5275356 | ||
BC066917.1 | - | 6 | 450 | 4792462 | ||
BG613555.1 | - | 6 | 545 | |||
BM541512.1 | - | 6 | 701 | |||
BQ428985.1 | - | 6 | 414 | |||
BU567649.1 | - | 6 | 589 | |||
BU570661.1 | - | 5 | 364 | |||
CR456916.1 | - | 5 | 254 | |||
DA335600.1 | - | 6 | 497 | |||
DA550074.1 | - | 6 | 351 | |||
DA965437.1 | - | 5 | 415 | |||
DB234695.1 | - | 5 | 353 | |||
U39318.1 | - | 5 | 299 |