Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003357.4 | NP_003348.1 | 1 | 91 | ||
GenBank mRNA | AK312165.1 | - | 1 | 91 | ||
BC004481.2 | - | 1 | 75 | 3688615 | ||
CR541780.1 | - | 1 | 30 | |||
DB196705.1 | - | 1 | 91 | |||
KU178373.1 | - | 1 | 30 | |||
KU178374.1 | - | 1 | 30 | |||
U01101.1 | - | 1 | 39 | |||
X13197.1 | - | 1 | 37 |