Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000551.3 | NP_000542.1 | 3 | 757 | ||
NM_198156.2 | NP_937799.1 | 2 | 634 | |||
GenBank mRNA | AK304002.1 | - | 3 | 342 | ||
AK309560.1 | - | 3 | 681 | |||
AK315799.1 | - | 3 | 616 | |||
BC058831.1 | - | 2 | 485 | 6457060 | ||
BX100230.1 | - | 1 | 218 | |||
KU178386.1 | - | 2 | 421 | |||
KU178387.1 | - | 3 | 544 | |||
L15409.1 | - | 3 | 757 |