Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 2, 3 | 461 | ||
NM_032463.2 | NP_115852.1 | 3, 4 | 580 | |||
NM_032464.2 | NP_115853.2 | 3, 4 | 650 | |||
XM_011516558.2 | XP_011514860.1 | 3, 4 | 490 | |||
GenBank mRNA | AF161531.1 | - | 2, 3 | 213 | ||
AF252611.1 | - | 3, 4 | 401 | |||
AF252612.1 | - | 2, 3 | 212 | |||
AF252613.1 | - | 2, 3 | 197 | |||
AF252614.1 | - | 3, 4 | 331 | |||
AF257135.1 | - | 1, 2 | 132 | |||
AK002099.1 | - | 3, 4 | 440 | |||
AK092904.1 | - | 3, 4 | 791 | |||
AK225150.1 | - | 2, 3 | 212 | |||
AK290074.1 | - | 3, 4 | 328 | |||
AK290916.1 | - | 2, 3 | 209 | |||
AY190023.1 | - | 1, 2 | 142 | |||
BC001609.1 | - | 3, 4 | 462 | 3543310 | ||
BC009204.2 | - | 1, 2 | 134 | 4054137 |