Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 10 | 780 | ||
NM_032463.2 | NP_115852.1 | 11 | 899 | |||
NM_032464.2 | NP_115853.2 | 11 | 969 | |||
XM_011516558.2 | XP_011514860.1 | 11 | 809 | |||
GenBank mRNA | AF161531.1 | - | 10 | 529 | ||
AF252611.1 | - | 11 | 720 | |||
AF252612.1 | - | 9 | 475 | |||
AF252613.1 | - | 10 | 516 | |||
AF252614.1 | - | 11 | 650 | |||
AF257135.1 | - | 9 | 451 | |||
AK002099.1 | - | 11 | 759 | |||
AK092904.1 | - | 11 | 1110 | |||
AK225150.1 | - | 10 | 531 | |||
AK290074.1 | - | 11 | 647 | |||
AK290916.1 | - | 10 | 528 | |||
AY190023.1 | - | 9 | 461 | |||
BC001609.1 | - | 11 | 781 | 3543310 | ||
BC009204.2 | - | 9 | 453 | 4054137 |