Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297560.1 | NP_001284489.1 | 3 | 477 | ||
NM_001297561.1 | NP_001284490.1 | 2 | 268 | |||
NM_021269.2 | NP_067092.2 | 3 | 393 | |||
XM_017027201.1 | XP_016882690.1 | 3 | 295 | |||
XM_017027202.1 | XP_016882691.1 | 2 | 294 | |||
XM_017027204.1 | XP_016882693.1 | 3 | 210 | |||
GenBank mRNA | AK128184.1 | - | 3 | 352 | ||
AK131527.1 | - | 3 | 393 | |||
BX647526.1 | - | 3 | 463 | |||
CD704852.1 | - | 2 | 290 | |||
DA811084.1 | - | 3 | 423 | |||
DB216212.1 | - | 2 | 266 | |||
X59845.1 | - | 2 | 159 |