Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006969.3 | NP_008900.3 | 4 | 3218 | ||
NR_036599.1 | 5 | 3336 | ||||
NR_036600.1 | 3 | 3091 | ||||
XM_006723354.3 | XP_006723417.1 | 4 | 3193 | |||
XM_011527262.2 | XP_011525564.1 | 4 | 3248 | |||
GenBank mRNA | AK091553.1 | - | 1 | 1126 | ||
AK092385.1 | - | 1 | 2025 | |||
AK131368.1 | - | 2 | 3816 | |||
AK131574.1 | - | 1 | 2603 | |||
AK304704.1 | - | 4 | 3182 | |||
BC140720.1 | - | 3 | 3119 | 9021588 | ||
BX640767.1 | - | 1 | 2246 | |||
CR933598.1 | - | 4 | 3181 |