Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145914.2 | NP_666019.1 | 3, 4 | 725 | ||
XM_005250568.4 | XP_005250625.1 | 4, 5 | 1867 | |||
XM_006716113.3 | XP_006716176.1 | 3, 4 | 1706 | |||
XM_017012584.1 | XP_016868073.1 | 4, 5 | 1831 | |||
XM_017012585.1 | XP_016868074.1 | 3, 4 | 749 | |||
GenBank mRNA | AF155100.1 | - | 2, 3 | 400 | ||
AK075324.1 | - | 4, 5 | 825 | |||
AK292267.1 | - | 3, 4 | 745 | |||
AK301871.1 | - | 5, 6 | 776 | |||
AK313884.1 | - | 3, 4 | 743 | |||
AL136865.1 | - | 2, 3 | 678 | |||
BC047309.1 | - | 3, 4 | 718 | 5111890 | ||
CR749301.1 | - | 4, 5 | 609 |