Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321163.1 | NP_001308092.1 | 4, 5 | 618 | ||
NM_021915.3 | NP_068734.1 | 4, 5 | 576 | |||
XM_006722874.3 | XP_006722937.1 | 4, 5 | 638 | |||
GenBank mRNA | BC051309.1 | - | 4, 5 | 505 | 5812091 |