Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321163.1 | NP_001308092.1 | 1 | 154 | ||
NM_021915.3 | NP_068734.1 | 1 | 154 | |||
XM_006722873.3 | XP_006722936.1 | 1 | 165 | |||
XM_006722874.3 | XP_006722937.1 | 1 | 165 | |||
XM_017027231.1 | XP_016882720.1 | 1 | 165 | |||
GenBank mRNA | BC051309.1 | - | 1 | 83 | 5812091 | |
BG393583.1 | - | 1 | 154 | |||
BG429615.1 | - | 1 | 110 |