Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256523.1 | NP_001243452.1 | 4 | 1295 | ||
NM_001256524.1 | NP_001243453.1 | 6 | 1153 | |||
NM_001256525.1 | NP_001243454.1 | 5 | 1053 | |||
NM_003426.3 | NP_003417.2 | 5 | 1381 | |||
NR_046282.1 | 6 | 1477 | ||||
GenBank mRNA | AB209663.1 | - | 6 | 1189 | ||
AK128267.1 | - | 2 | 2386 | |||
AK291358.1 | - | 6 | 1079 | |||
AK299569.1 | - | 5 | 979 | |||
BC013395.2 | - | 5 | 979 | 3028001 | ||
BC048295.1 | - | 5 | 966 | 5242841 | ||
CR456616.1 | - | 5 | 920 | |||
X71623.1 | - | 4 | 813 | |||
X92715.1 | - | 4 | 1979 |