Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF204P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002722.2 | 4 | 3613 | |||
NR_024553.1 | 5 | 2493 | ||||
GenBank mRNA | AK296042.1 | - | 1 | 1217 | ||
AL833520.1 | - | 4 | 3614 | |||
BC067282.1 | - | 2 | 2465 |