Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321645.1 | NP_001308574.1 | 6 | 2450 | ||
NM_013398.3 | NP_037530.2 | 6 | 2408 | |||
XM_017027261.1 | XP_016882750.1 | 5 | 2484 | |||
GenBank mRNA | AF067164.1 | - | 1 | 2925 | ||
AF187990.2 | - | 6 | 2393 | |||
AK292500.1 | - | 6 | 2388 | |||
AY148489.1 | - | 1 | 1836 | |||
BC051902.1 | - | 6 | 2345 | 6155240 |
Gene Symbol: LOC100379224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033341.1 | 2 | 445 | |||
GenBank mRNA | BC044807.1 | - | 2 | 407 | ||
BC047357.1 | - | 2 | 407 |