Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270399.1 | NP_001257328.1 | 4 | 1567 | ||
NM_001270400.1 | NP_001257329.1 | 4 | 1095 | |||
NM_006009.3 | NP_006000.2 | 4 | 1007 | |||
GenBank mRNA | AF141347.1 | - | 4 | 741 | ||
AF141348.1 | - | 1 | 294 | |||
AK057842.1 | - | 4 | 761 | |||
AK289483.1 | - | 4 | 934 | |||
AK298838.1 | - | 4 | 617 | |||
BC006468.1 | - | 4 | 716 | 3528520 | ||
BC050637.2 | - | 4 | 746 | 6050536 | ||
K00557.1 | - | 2 | 335 |