Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005435.3 | NP_005426.2 | 4 | 3350 | ||
XM_017012623.1 | XP_016868112.1 | 4 | 3405 | |||
GenBank mRNA | AK090570.1 | - | 3 | 993 | ||
AK123596.1 | - | 3 | 1549 | |||
AK160365.1 | - | 4 | 3406 | |||
BC010046.2 | - | 4 | 295 | |||
BC011839.2 | - | 4 | 295 | |||
BC014555.2 | - | 4 | 295 | |||
BC136661.1 | - | 4 | 3264 | 9020651 | ||
BX649178.1 | - | 3 | 1235 | |||
CR749203.1 | - | 4 | 3290 | |||
U02082.1 | - | 2 | 91 |
Gene Symbol: ARHGEF34P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033942.1 | 3 | 2269 |