Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005435.3 | NP_005426.2 | 14 | 4666 | ||
GenBank mRNA | AK090570.1 | - | 13 | 2116 | ||
AK123596.1 | - | 13 | 2865 | |||
AK160365.1 | - | 14 | 4722 | |||
BC010046.2 | - | 14 | 1611 | |||
BC011839.2 | - | 14 | 1611 | |||
BC014555.2 | - | 14 | 1611 | |||
BC136661.1 | - | 14 | 4580 | 9020651 | ||
BX649178.1 | - | 13 | 2551 | |||
U02082.1 | - | 12 | 1407 |