Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014564.4 | NP_055379.1 | 3 | 542 | ||
NM_178138.5 | NP_835258.1 | 3 | 510 | |||
XM_005263410.1 | XP_005263467.1 | 3 | 824 | |||
XM_017015168.1 | XP_016870657.1 | 2 | 321 | |||
GenBank mRNA | AB593062.1 | - | 3 | 510 | ||
AB593063.1 | - | 3 | 509 | |||
AB593064.1 | - | 3 | 824 | |||
AB593065.1 | - | 2 | 1218 | |||
AB593066.1 | - | 3 | 542 | |||
AB593067.1 | - | 3 | 506 | |||
AF096169.1 | - | 2 | 314 | |||
AF156888.1 | - | 3 | 497 | |||
AF156889.1 | - | 3 | 527 |