Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005088.2 | NP_005079.2 | 5 | 2270 | ||
NR_027383.1 | 6 | 2340 | ||||
GenBank mRNA | BC110496.1 | - | 5 | 2268 | 40007662 | |
BC110497.1 | - | 5 | 2268 | 40007663 | ||
L03426.1 | - | 6 | 2310 | |||
M99578.1 | - | 5 | 2241 |