Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003498.5 | NP_003489.1 | 2 | 374 | ||
XM_017023741.1 | XP_016879230.1 | 2 | 402 | |||
XM_017023742.1 | XP_016879231.1 | 2 | 300 | |||
GenBank mRNA | AF030196.1 | - | 2 | 326 | ||
AF070673.1 | - | 2 | 356 | |||
AL161976.1 | - | 2 | 335 | |||
BC036100.1 | - | 2 | 352 | 5277179 | ||
BC036443.1 | - | 2 | 346 | 4823944 | ||
CR542127.1 | - | 1 | 181 | |||
CR542138.1 | - | 1 | 181 | |||
CR984360.1 | - | 2 | 362 |