Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004182.3 | NP_004173.1 | 3, 4 | 265 | ||
NM_153477.2 | NP_705582.1 | 2, 3 | 443 | |||
NR_045560.1 | 3, 4 | 265 | ||||
GenBank mRNA | AF083241.1 | - | 3, 4 | 231 | ||
AF083242.1 | - | 2, 3 | 381 | |||
AF092737.1 | - | 3, 4 | 246 | |||
AK223320.1 | - | 3, 4 | 219 | |||
AK312072.1 | - | 3, 4 | 231 | |||
BC000720.1 | - | 2, 3 | 200 | 3505415 | ||
BC008890.1 | - | 2, 3 | 200 | 3959862 | ||
DA372561.1 | - | 3, 4 | 265 | |||
EL949569.1 | - | 3, 4 | 243 |