Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002174.2 | 3 | 372 | |||
NR_027626.1 | 4 | 691 | ||||
GenBank mRNA | AF074480.1 | - | 4 | 392 | ||
AK303297.1 | - | 4 | 648 | |||
BC032500.1 | - | 4 | 584 | |||
CR749466.1 | - | 2 | 175 | |||
FJ794466.1 | - | 3 | 350 |