Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002174.2 | 6 | 794 | |||
NR_027626.1 | 7 | 1113 | ||||
GenBank mRNA | AF074480.1 | - | 7 | 814 | ||
AK303297.1 | - | 7 | 1070 | |||
BC032500.1 | - | 7 | 1006 | |||
CR749466.1 | - | 5 | 597 |