Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003614.1 | NP_003605.1 | 1 | 319 | ||
GenBank mRNA | AF067733.1 | - | 1 | 294 | ||
AF073799.1 | - | 1 | 319 | |||
AY587582.1 | - | 1 | 294 |
Set Membership: |
G-protein coupled receptor |