Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003693.3 | NP_003684.2 | 11 | 2445 | ||
NM_145350.2 | NP_663325.1 | 11 | 2408 | |||
NR_028075.2 | 11 | 2375 | ||||
NR_102409.1 | 11 | 2482 | ||||
XM_017025256.1 | XP_016880745.1 | 7 | 1346 | |||
GenBank mRNA | AB052947.1 | - | 11 | 2309 | ||
AB052948.1 | - | 11 | 2375 | |||
AB052949.1 | - | 10 | 2784 | |||
AB052950.1 | - | 9 | 2111 | |||
AK291810.1 | - | 11 | 2388 | |||
AK298737.1 | - | 8 | 1527 | |||
AK310174.1 | - | 11 | 2405 | |||
BC039735.1 | - | 11 | 2413 | 6041360 | ||
D63483.1 | - | 11 | 2355 | |||
D86864.1 | - | 11 | 2379 |