Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320296.1 | NP_001307225.1 | 1 | 533 | ||
NM_003717.3 | NP_003708.1 | 2 | 303 | |||
GenBank mRNA | AF005271.1 | - | 2 | 302 | ||
BC104234.1 | - | 1 | 482 | 40033026 | ||
BC104235.1 | - | 1 | 482 | 40033028 |