Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFXANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278727.1 | NP_001265656.1 | 6, 7 | 1012 | ||
NM_001278728.1 | NP_001265657.1 | 5, 6 | 847 | |||
NM_003721.3 | NP_003712.1 | 7, 8 | 1078 | |||
NM_134440.2 | NP_604389.1 | 6, 7 | 1009 | |||
XM_005260134.4 | XP_005260191.1 | 7, 8 | 740 | |||
XM_005260135.3 | XP_005260192.1 | 7, 8 | 740 | |||
XM_005260136.4 | XP_005260193.1 | 7, 8 | 1039 | |||
XM_005260137.4 | XP_005260194.1 | 7, 8 | 810 | |||
XM_006722930.3 | XP_006722993.1 | 6, 7 | 908 | |||
XM_017027415.1 | XP_016882904.1 | 6, 7 | 901 | |||
XM_017027416.1 | XP_016882905.1 | 5, 6 | 835 | |||
GenBank mRNA | AF077196.1 | - | 6, 7 | 727 | ||
AF094760.1 | - | 7, 8 | 990 | |||
AF105427.1 | - | 6, 7 | 735 | |||
AF105428.1 | - | 5, 6 | 649 | |||
AW411190.1 | - | 3, 4 | 374 | |||
AW411361.1 | - | 3, 4 | 382 | |||
BC114558.1 | - | 6, 7 | 968 | 40032318 | ||
BC114563.1 | - | 7, 8 | 1034 | 40032319 | ||
BG740813.1 | - | 5, 6 | 702 | |||
CR456981.1 | - | 5, 6 | 573 | |||
CR542199.1 | - | 5, 6 | 573 |