Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFXANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278727.1 | NP_001265656.1 | 3 | 665 | ||
NM_001278728.1 | NP_001265657.1 | 2 | 503 | |||
NM_003721.3 | NP_003712.1 | 3 | 665 | |||
NM_134440.2 | NP_604389.1 | 3 | 665 | |||
XM_005260134.4 | XP_005260191.1 | 3 | 327 | |||
XM_005260135.3 | XP_005260192.1 | 3 | 327 | |||
XM_005260136.4 | XP_005260193.1 | 3 | 629 | |||
XM_005260137.4 | XP_005260194.1 | 3 | 400 | |||
XM_006722930.3 | XP_006722993.1 | 2 | 498 | |||
XM_017027415.1 | XP_016882904.1 | 2 | 488 | |||
XM_017027416.1 | XP_016882905.1 | 2 | 488 | |||
GenBank mRNA | AF077196.1 | - | 2 | 314 | ||
AF094760.1 | - | 3 | 577 | |||
AF105427.1 | - | 2 | 322 | |||
AF105428.1 | - | 2 | 305 | |||
BC114558.1 | - | 3 | 621 | 40032318 | ||
BC114563.1 | - | 3 | 621 | 40032319 | ||
BG740813.1 | - | 2 | 358 | |||
BM712354.1 | - | 2 | 503 | |||
CR456981.1 | - | 1 | 160 | |||
CR542199.1 | - | 1 | 160 | |||
CX871218.1 | - | 3 | 598 |