Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003745.1 | NP_003736.1 | 2 | 173 | ||
GenBank mRNA | AB000734.1 | - | 2 | 173 | ||
AB005043.1 | - | 1 | 72 | |||
AK127621.1 | - | 1 | 741 | |||
U88326.1 | - | 1 | 42 |