Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284336.1 | NP_001271265.1 | 4 | 420 | ||
NM_003758.3 | NP_003749.2 | 6 | 567 | |||
GenBank mRNA | AF090923.1 | - | 2 | 163 | ||
AK002006.1 | - | 6 | 487 | |||
AK023388.1 | - | 6 | 517 | |||
BC002719.2 | - | 6 | 500 | 3632801 | ||
U97670.1 | - | 6 | 500 |