Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320099.1 | NP_001307028.1 | 2 | 523 | ||
NM_001320100.1 | NP_001307029.1 | 2 | 313 | |||
NM_001320101.1 | NP_001307030.1 | 2 | 313 | |||
NM_003805.4 | NP_003796.1 | 2 | 313 | |||
NR_135147.1 | 2 | 313 | ||||
XM_017020142.1 | XP_016875631.1 | 2 | 342 | |||
XM_017020143.1 | XP_016875632.1 | 2 | 336 | |||
XM_017020144.1 | XP_016875633.1 | 2 | 336 | |||
XM_017020145.1 | XP_016875634.1 | 2 | 335 | |||
GenBank mRNA | AB451280.1 | - | 1 | 205 | ||
AB451412.1 | - | 1 | 205 | |||
AI791547.1 | - | 2 | 307 | |||
AK294986.1 | - | 2 | 301 | |||
AK296225.1 | - | 2 | 311 | |||
AK314861.1 | - | 2 | 293 | |||
BC017042.1 | - | 2 | 285 | 3849848 | ||
BC037905.1 | - | 2 | 527 | 5276510 | ||
BT009837.1 | - | 1 | 205 | |||
CR407643.1 | - | 1 | 205 | |||
HY113325.1 | - | 2 | 314 | |||
U79115.1 | - | 2 | 305 | |||
U84388.1 | - | 2 | 242 |