Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004195.2 | NP_004186.1 | 3 | 515 | ||
NM_148901.1 | NP_683699.1 | 3 | 515 | |||
NM_148902.1 | NP_683700.1 | 3 | 515 | |||
XM_017002722.1 | XP_016858211.1 | 3 | 1706 | |||
GenBank mRNA | AF117297.1 | - | 3 | 377 | ||
AF125304.1 | - | 3 | 377 | |||
AF241229.1 | - | 3 | 377 | |||
AY358877.1 | - | 3 | 497 | |||
BC152381.1 | - | 3 | 380 | 100013440 | ||
BC152386.1 | - | 3 | 380 | |||
BT019531.1 | - | 3 | 377 | |||
BT019532.1 | - | 3 | 377 | |||
KP067911.1 | - | 3 | 515 |