Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003848.3 | NP_003839.2 | 11 | 1270 | ||
XM_017007420.1 | XP_016862909.1 | 11 | 1912 | |||
GenBank mRNA | AF058954.1 | - | 10 | 1158 | ||
AF131748.1 | - | 5 | 543 | |||
AL050226.1 | - | 5 | 531 | |||
BC007716.2 | - | 11 | 1233 | 4303307 | ||
BC019868.1 | - | 8 | 909 | |||
BC035149.1 | - | 10 | 1032 | |||
BC047024.1 | - | 12 | 1240 | 5415743 | ||
BC068602.1 | - | 11 | 1271 | 5274341 |