Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004706.3 | NP_004697.2 | 13 | 1174 | ||
NM_198977.1 | NP_945328.1 | 12 | 1075 | |||
NM_199002.1 | NP_945353.1 | 13 | 1199 | |||
XM_005259386.4 | XP_005259443.1 | 13 | 1526 | |||
XM_005259390.4 | XP_005259447.1 | 13 | 1522 | |||
XM_011527468.2 | XP_011525770.1 | 13 | 1522 | |||
GenBank mRNA | AL832294.1 | - | 2 | 158 | ||
BC005155.2 | - | 10 | 912 | |||
BC011726.2 | - | 12 | 1050 | 4138058 | ||
BC034013.1 | - | 13 | 1130 | 3451036 | ||
BC067262.1 | - | 12 | 1018 | 6502165 | ||
BT007421.1 | - | 10 | 888 | |||
U64105.1 | - | 13 | 1119 |