Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004706.3 | NP_004697.2 | 15, 16 | 1534 | ||
NM_198977.1 | NP_945328.1 | 14, 15 | 1435 | |||
NM_199002.1 | NP_945353.1 | 15, 16 | 1559 | |||
XM_005259386.4 | XP_005259443.1 | 17, 18 | 2066 | |||
XM_005259390.4 | XP_005259447.1 | 15, 16 | 1882 | |||
XM_011527468.2 | XP_011525770.1 | 16, 17 | 1894 | |||
GenBank mRNA | AL832294.1 | - | 6, 7 | 698 | ||
BC005155.2 | - | 12, 13 | 1272 | |||
BC011726.2 | - | 14, 15 | 1410 | 4138058 | ||
BC034013.1 | - | 15, 16 | 1490 | 3451036 | ||
BC035127.1 | - | 2, 3 | 1072 | |||
BC067262.1 | - | 14, 15 | 1378 | 6502165 | ||
BT007421.1 | - | 12, 13 | 1248 | |||
U64105.1 | - | 15, 16 | 1479 |