Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004708.3 | NP_004699.1 | 3, 4 | 248 | ||
XM_005259392.4 | XP_005259449.1 | 3, 4 | 275 | |||
GenBank mRNA | AF014955.1 | - | 3, 4 | 198 | ||
AK293486.1 | - | 3, 4 | 246 | |||
BC015519.2 | - | 3, 4 | 214 | 3888459 | ||
BM696336.1 | - | 3, 4 | 238 | |||
BT006694.1 | - | 3, 4 | 174 | |||
CR456934.1 | - | 3, 4 | 174 | |||
DN915905.1 | - | 3, 4 | 248 |