Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004709.2 | NP_004700.1 | 1 | 1213 | ||
GenBank mRNA | AF007148.1 | - | 1 | 664 | ||
Y08902.1 | - | 1 | 1213 |
Gene Symbol: SLITRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005262342.3 | XP_005262399.1 | 5 | 7789 | ||
XM_005262343.3 | XP_005262400.1 | 5 | 7799 | |||
XM_005262344.3 | XP_005262401.1 | 5 | 7777 | |||
XM_005262345.3 | XP_005262402.1 | 4 | 7640 |