Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 8 | 1189 | ||
GenBank mRNA | AK299293.1 | - | 8 | 1148 | ||
AK301461.1 | - | 7 | 957 | |||
AK310703.1 | - | 8 | 1130 | |||
AK310938.1 | - | 6 | 689 | |||
AL833263.1 | - | 7 | 912 | |||
BC033903.1 | - | 8 | 1171 | |||
BC107880.1 | - | 8 | 1170 |