Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004228.6 | NP_004219.3 | 4 | 590 | ||
NM_017457.5 | NP_059431.1 | 4 | 590 | |||
XM_006723472.2 | XP_006723535.1 | 4 | 676 | |||
XM_006723473.2 | XP_006723536.1 | 4 | 676 | |||
GenBank mRNA | AB209573.1 | - | 4 | 422 | ||
AK292405.1 | - | 4 | 447 | |||
AK299590.1 | - | 4 | 433 | |||
BC004361.1 | - | 4 | 405 | 3538580 | ||
BC038713.1 | - | 4 | 447 | 5269719 | ||
U70728.1 | - | 4 | 448 | |||
X99753.1 | - | 4 | 290 |