Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301061.1 | NP_001287990.1 | 5 | 568 | ||
NM_001707.3 | NP_001698.2 | 4 | 446 | |||
NR_036682.1 | 4 | 429 | ||||
GenBank mRNA | AJ223979.1 | - | 4 | 325 | ||
AK290091.1 | - | 4 | 354 | |||
AL525884.3 | - | 5 | 509 | |||
AL553260.3 | - | 4 | 713 | |||
BC000956.2 | - | 4 | 405 | 3450946 | ||
BC001967.2 | - | 4 | 312 | 3461829 | ||
BC009548.1 | - | 4 | 356 | 3897805 | ||
BI603803.1 | - | 4 | 442 | |||
BQ109412.1 | - | 4 | 378 | |||
DA688517.1 | - | 4 | 429 | |||
X89985.1 | - | 4 | 326 |