Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
siRNA ID
HSS145090
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
8
1216
NM_001145771.2
NP_001139243.1
8
1654
NM_001145772.2
NP_001139244.1
8
1680
NM_001145773.2
NP_001139245.1
7
1551
NM_001145774.2
NP_001139246.1
8
1453
NM_001290142.1
NP_001277071.1
7
756
NM_001290143.1
NP_001277072.1
7
1105
NM_001290144.1
NP_001277073.1
7
1292
NM_005682.6
NP_005673.3
8
1216
NM_201524.3
NP_958932.1
8
1658
NM_201525.3
NP_958933.1
7
1536
XM_005256237.2
XP_005256294.1
7
1252
XM_005256238.2
XP_005256295.1
8
1591
XM_005256239.2
XP_005256296.1
8
1353
XM_005256240.4
XP_005256297.1
7
1074
XM_005256241.4
XP_005256298.1
7
1078
XM_005256242.4
XP_005256299.1
7
1135
XM_005256244.3
XP_005256301.1
7
1227
XM_005256245.2
XP_005256302.1
7
1278
XM_005256246.2
XP_005256303.1
8
1338
XM_005256247.2
XP_005256304.1
7
1237
XM_005256248.2
XP_005256305.1
7
1302
XM_005256249.3
XP_005256306.1
7
1093
XM_005256251.4
XP_005256308.1
7
1119
XM_005256252.2
XP_005256309.1
7
1063
XM_005256254.2
XP_005256311.1
7
1263
XM_005256255.2
XP_005256312.1
8
1576
XM_006721338.2
XP_006721401.1
8
1374
XM_006721339.3
XP_006721402.1
8
1181
XM_006721340.3
XP_006721403.1
8
1439
XM_006721341.2
XP_006721404.1
8
1396
XM_006721342.2
XP_006721405.1
8
1359
XM_006721343.3
XP_006721406.1
8
1424
XM_006721344.2
XP_006721407.1
8
1381
XM_006721345.3
XP_006721408.1
8
1150
XM_006721346.2
XP_006721409.1
7
1005
XM_006721347.2
XP_006721410.1
8
1591
XM_011523461.2
XP_011521763.1
8
1342
XM_011523462.2
XP_011521764.1
7
1157
XM_011523463.2
XP_011521765.1
8
1167
XM_011523464.2
XP_011521766.1
8
1327
XM_011523465.2
XP_011521767.1
8
1265
XM_011523466.2
XP_011521768.1
7
1312
XM_011523467.2
XP_011521769.1
8
1059
XM_011523468.2
XP_011521770.1
7
1061
XM_017023892.1
XP_016879381.1
7
1263
GenBank mRNA
AF106858.1
-
7
1086
AK075270.1
-
8
1168
AK297236.1
-
7
1165
AK312405.1
-
7
1372
AY358400.1
-
8
1137
BC013207.1
-
8
1230
EU432119.1
-
6
924
KU178472.1
-
6
924
KU178473.1
-
6
924
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline