Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRSF11 SIRNA Assays ›
Gene Symbol:
SRSF11
Gene Name:
serine and arginine rich splicing factor 11
Gene Aliases:
NET2, SFRS11, dJ677H15.2, p54
Chromosome Location:
Chr.1: 70205682 - 70252020 on Build GRCh38
UniGene ID:
Hs.479693
Species:
Human
siRNA ID
HSS145093
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SRSF11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190987.1
NP_001177916.1
13
1511
NM_004768.3
NP_004759.1
13
1517
XM_005271338.3
XP_005271395.1
13
1529
XM_005271339.3
XP_005271396.1
13
1531
XM_011542427.2
XP_011540729.1
12
1670
XM_011542428.2
XP_011540730.1
12
4927
XM_011542431.2
XP_011540733.1
13
2691
XM_017002811.1
XP_016858300.1
13
1526
XM_017002812.1
XP_016858301.1
13
1508
XM_017002813.1
XP_016858302.1
13
1507
XM_017002814.1
XP_016858303.1
11
2660
XM_017002815.1
XP_016858304.1
13
4272
XM_017002816.1
XP_016858305.1
14
4406
XM_017002817.1
XP_016858306.1
14
3980
XM_017002818.1
XP_016858307.1
13
1808
XM_017002819.1
XP_016858308.1
14
1917
XM_017002820.1
XP_016858309.1
13
4632
XM_017002821.1
XP_016858310.1
13
1805
XM_017002822.1
XP_016858311.1
11
3002
XM_017002823.1
XP_016858312.1
12
2347
XM_017002824.1
XP_016858313.1
14
4385
XM_017002825.1
XP_016858314.1
14
3959
XM_017002826.1
XP_016858315.1
13
1787
XM_017002827.1
XP_016858316.1
14
1896
XM_017002828.1
XP_016858317.1
15
3730
XM_017002829.1
XP_016858318.1
15
3766
XM_017002830.1
XP_016858319.1
13
4888
XM_017002831.1
XP_016858320.1
14
3956
XM_017002832.1
XP_016858321.1
13
1784
XM_017002833.1
XP_016858322.1
14
1893
XM_017002834.1
XP_016858323.1
12
2626
GenBank mRNA
AL832279.1
-
10
1776
BC040436.1
-
13
1469
5518580
BX640645.1
-
11
2754
BX641004.1
-
9
937
M74002.1
-
13
1476
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA processing
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0