Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004813.2 | NP_004804.1 | 1 | 349 | ||
NM_057174.2 | NP_476515.1 | 1 | 349 | |||
GenBank mRNA | AB016531.1 | - | 1 | 74 | ||
AF118240.1 | - | 1 | 281 | |||
BC000467.2 | - | 1 | 55 | 2822827 | ||
BC004356.1 | - | 1 | 47 | 2822311 | ||
CN285513.1 | - | 1 | 356 |