Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033677.1 | NP_001028849.1 | 3, 4 | 896 | ||
NM_004276.4 | NP_004267.2 | 3, 4 | 351 | |||
NM_031205.3 | NP_112482.1 | 4, 5 | 531 | |||
XM_017020235.1 | XP_016875724.1 | 3, 4 | 963 | |||
XM_017020236.1 | XP_016875725.1 | 3, 4 | 1098 | |||
XM_017020237.1 | XP_016875726.1 | 3, 4 | 1098 | |||
XM_017020238.1 | XP_016875727.1 | 4, 5 | 2177 | |||
XM_017020239.1 | XP_016875728.1 | 3, 4 | 1991 | |||
GenBank mRNA | AF169148.1 | - | 3, 4 | 290 | ||
AF169149.1 | - | 4, 5 | 470 | |||
AK127203.1 | - | 1, 2 | 1365 | |||
BC030201.1 | - | 3, 4 | 896 | 5200625 |