Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323408.1 | NP_001310337.1 | 1 | 200 | ||
NM_001323409.1 | NP_001310338.1 | 1 | 200 | |||
NM_001323410.1 | NP_001310339.1 | 1 | 200 | |||
NM_006814.4 | NP_006805.2 | 1 | 200 | |||
NM_178578.3 | NP_848693.2 | 2 | 221 | |||
GenBank mRNA | AK098612.1 | - | 1 | 166 | ||
AK294134.1 | - | 1 | 156 | |||
BC126462.1 | - | 2 | 77 | 8992178 | ||
BI461198.1 | - | 1 | 196 | |||
BI830805.1 | - | 2 | 154 | |||
D88378.1 | - | 1 | 145 | |||
DA949012.1 | - | 1 | 157 | |||
DB028624.1 | - | 2 | 187 | |||
DC398668.1 | - | 1 | 200 |