Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323409.1 | NP_001310338.1 | 7 | 997 | ||
NM_006814.4 | NP_006805.2 | 7 | 981 | |||
NM_178578.3 | NP_848693.2 | 8 | 1002 | |||
GenBank mRNA | AK300667.1 | - | 7 | 726 | ||
BC029836.1 | - | 4 | 716 | |||
BC126462.1 | - | 8 | 858 | 8992178 | ||
D88378.1 | - | 7 | 926 |